Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius™ MGG Grade Glass Microfiber Filters w/o Binder

Manufacturer:  Sartorius™ FT31106025

 View more versions of this product

Catalog No. 14-555-130


Add to cart

Description

Description

Ideal for critical analyses in which impurities can be washed out of the filter.
Specifications

Specifications

32mbar
0.98 in.
Filter Disc
1.5μm
932°F
0.01 in.
2.5cm
Borosilicate Microfiber Glass
White
38 sec.
MGG
Non-sterile
500°C
0.28mm
Ideal for critical analyses in which impurities can be washed out of the filter
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit