Learn More
Sartorius™ Microsart™ e.motion Membrane Filters
For colony counting, particle testing and microscopy. Specifically designed for use in the Microsart™ e.motion dispenser, the filters are individually sterile-sealed in a format packaged on a pleated band, without protective paper. Can also be used with Sartorius nutrient pad sets.
Manufacturer:  Sartorius™ 139H6Z47SCM
Catalog No. 14-555-658
Add to cart
Description
Description
Microsart™ e.motion Membrane Filters, 0.45µm HIGH FLOW, white/green,47 mm The Microsart™ e.motion membrane filters are suitable for the use with the Microsart™ e.motion dispenser. The filters are individually sterile-sealed on a band without any protective paper. The special pleating as well as the shape of the sealing of the band of membrane filters ensures a perfectly flat and uniform dispensing of the individual membrane filters. The Microsart™ e.motion membrane filters are also ideal for all users of the Sartorius Nutrient Pad Sets: most of the NPS types are available with Microsart™ e.motion membrane filters. Outstanding recovery rates for microorganisms 0.45 µm are acc. to ISO 7704. Multi-fit: Fits into other dispensers. Protective paper-free. Packaged on a special pleated band. Product data is printed on individual envelopes. High flow membranes. Gamma irradiated, 25 kGray.
Specifications
Specifications
Sterile
Cellulose Nitrate
White
For colony count; sterility testing; particle count; microscopy
0.45μm
47mm
300/pk.
Green
Documents
Promotions
Promotions
  • Sartorius , Fisherbrand , Fisher Scientific
    Buy $1,500 or more in Sartorius products and get a Sartorius Tacta Pipette Multipack at no additional cost.
    06-30-2021