Learn More
Sartorius™ Microsart™ e.motion Membrane Filters Available on GSA/VA Contract for Federal Government customers only.
Membrane Filters suitable for use in the Microsart e.motion dispenser
Manufacturer:  Sartorius™ 13906Z50SCM
 View more versions of this product
Catalog No. 5090117784
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
0.45μm
50mm
Yes
For colony count; sterility testing; particle count; microscopy
Cellulose Nitrate
White
300/pk.
Green