Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius™ Midrange pHBasic+ Series Benchtop Meters

Complete functionality at your fingertips!

Manufacturer:  Sartorius™ PHBASIC+BIOKIT/DEL

 View more versions of this product

Catalog No. 14-557-879

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

  • Provides fast and accurate results
  • Easy-to-use menus and intuitive operation sets
  • Superior accuracy with up to 5-point Calibration with AutoCal feature
  • Large LCD display integrated in keyboard plastic foil
  • Sensor efficiency and calibration integrity with ElectCheck
  • Clear status icons and indicators like Stability Control
  • Automatic temperature compensation
  • Automatic buffer recognition
  • Programmable data collection
  • Recall and output of 500 data set memory provides convenient and easy way to manipulate pH results for analysis
  • Full functionality with USB data transmission
  • Automatic switch-over to manual temperature input when no temperature sensor is connected


Specifications:
  • Measuring Value Storage (manual/automatic): 500 data sets, storage intervals from 5 sec. to 60 min.
  • Lower Housing : Plastic
  • Plastic Foil Keypad with Tactile Response: yes
  • External Universal Power Supply: primary: 100 to 240V, 50/60Hz; secondary: 9V = 1.5A
  • Battery Type: automatic switch-off 4 mignon battery (adjustable 10 min. to 24 hr., default 1 h, cannot be switched off)
  • Climate Class: 2 (VDI/VDE 3540)
  • IQ/OQ Documents Available: Yes
  • Dimensions ( D x W x H): 24 x 19 x 8cm
  • Weight: approx.1kg


pH Measurements:
  • Range: -2.0 to +20.0, -2.00 to +20.00 , -2.000 to +19.999
  • Readability: 0.1, 0.01, 0.001
  • Accuracy: ±0.1, ±0.01, ±0.005
  • Pre-programed Buffer Sets: 16
  • Channels: One
  • Connector Type: BNC
  • Manually Enter Buffer: Yes
  • Reference Connector Type: 4mm banana plug


mV Measurements:
  • Range: -1200.0 to +1200.0, -2000 to +2000
  • Readability: 0.1, 1
  • Accuracy: ±0.3, ±1
  • Auto Range: Yes
  • Channels: One


Temperature Measurements:
  • Range: -5° to +105°
  • Readability: 0.1
  • Accuracy: ±0.1°C
  • Channels: One
  • Selectable °C/°F: Yes
  • Manually Adjustable: up to 130°C
  • Connector Types: 2.5mm phone jack connector for 10KOhm


Includes:
  • Cat. No.: 14-557-876 pHBasic+A Meter only: Meter with Power Supply, Cover
  • Cat. No. 14-557-877 pHBasic+ Meter Kit: Meter, Power Supply, Cover, Arm
  • Cat. No. 14-557-878 pHBasic+ pH Kit: Meter, Power Supply, Cover, Arm, three Buffers, Electrode: PY-P10
  • Cat. No. 14-557-879 pHBasic+ Bio Kit: Meter, Power Supply, Cover, Arm, three Buffers, Electrode: PY-P28
  • Cat. No. 14-557-880 pHBasic+ Food Kit: Meter, Power Supply, Cover, Arm, three Buffers, Electrode: PY-P30

Certifications

Protection class III, EG guidelines 73/23,EN 61010-1: 2001

Compliance

CE, cETLus

Warranty and Services

Two years.

Specifications

Specifications

500 Data Sets
Automatic
-5° to +105°
7.4 in.
19cm
±0.1°C
USB data transmission
24cm
External Universal Power Supply: primary: 120/240V, 50/60Hz, secondary: 9V = 1.5A
-2.0 to 20.0, -2.00 to 20.00, -2.000 to 19.999
Meter, Power Supply, Cover, Arm, three Buffers, PY-P28 Electrode
0.1, 1
TFT
0.1
approx.1kg
±0.1, ±0.01, ±0.005
±0.3, ±1
9.4 in.
Protection class III, EG guidelines 73/23,EN 61010-1: 2001
8cm
0.1, 0.01, 0.001
3 in.
Plastic
-1200.0 to 1200.0 mV, -2000 to 2000 mV
Documents
Promotions

Promotions

  • Sartorius , Fisher Scientific , Fisherbrand
    Buy $1,500 or more in Sartorius products and get a Sartorius Tacta Pipette Multipack at no additional cost.

    06-30-2021

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit