Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Sartorius™ Minisart™ RC Syringe Filters, Sterile Available on GSA/VA Contract for Federal Government customers only.
For rapid small volume particle removal filtration with maximum user convinience.
Manufacturer:  Sartorius™ 17764ACK
 View more versions of this product
Catalog No. 14-555-317
Description
Description
Minisart RC Units with hydrophilic, solvent-resistant RC-membranes. Ready-to-use syringe filter units for simple, rapid and reliable ultracleaning of small-volume samples for the HPLC or GC analysis. Minisart RC units outperform competitive hydrophilic units in terms of compatibility with aqueous solutions and solvent mixtures. Minisart Syringe Filter holders for rapid small volume particle removal filtration with maximum user convinience. Fitted on standard syringes.
  • Sample cleaning
  • Particle removal
  • Easy to use
  • Proven quality
  • 100 % tested during manufacture
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Syringe Filter
Regenerated Cellulose (RC)
0.2μm
25mm
Yes