Learn More
Sartorius™ Minisart™ RC Syringe Filters, Sterile
For rapid small volume particle removal filtration with maximum user convinience.
Manufacturer:  Sartorius™ 17761ACK
 View more versions of this product
Catalog No. 14-555-311
Add to cart
Description
Description
Minisart RC Units with hydrophilic, solvent-resistant RC-membranes. Ready-to-use syringe filter units for simple, rapid and reliable ultracleaning of small-volume samples for the HPLC or GC analysis. Minisart RC units outperform competitive hydrophilic units in terms of compatibility with aqueous solutions and solvent mixtures. Minisart Syringe Filter holders for rapid small volume particle removal filtration with maximum user convinience. Fitted on standard syringes.
  • Sample cleaning
  • Particle removal
  • Easy to use
  • Proven quality
  • 100 % tested during manufacture
Specifications
Specifications
Syringe Filter
Regenerated Cellulose (RC)
0.2μm
15mm
Yes