Learn More
Sartorius™ Minisart™ SRP4 Syringe Filters
Minisart™ SRP with chemically inert hydrophobic PTFE is suitable for venting as well as harsh solvents with a pH of 1 - 14. Type 17844 with a pore size of 0.2 µm can be used for sample preparation and sterile filtration. It has a male luer slip outlet. Version Q is a non-sterile bulk pack. Units can be sterilized by autoclaving or EO.
Manufacturer:  Sartorius™ 17844Q
Catalog No. 14-556-105
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • Low adsorption of analytes
  • Maximum chemical compatibility
  • Minimum extractables or leachables
  • Bidirectional use possible
  • 100% optical integrity test