Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius™ Nutrient Pad Sets (NPS) for Microsart™ e.motion Dispenser Available on GSA/VA Contract for Federal Government customers only.

Sterile, dehydrated culture media which is individually inserted in a petri dish and sterilized

Manufacturer:  Sartorius™ 1405650RDN

 View more versions of this product

Catalog No. 5090117643


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

DIN and ISO standards
1.96 in.
E. coli and coliforms
Green
0.45μm
Yes
White
5cm
Dehydrated
Individually packaged
Tergitol TTC

Safety and Handling

ShelfLife : 2 Years

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit