Sartorius™ PlatinumLINE Electrodes, pH

Get fast and accurate results with high quality materials, low-resistance A type glass membrane or unique Platinum junctions with patented Ag/AgCl reference system

Manufacturer: Sartorius™ PYP222S

 View more versions of this product

Catalog No. 14-557-225

  Check Availability
Add to Cart

    Specifications

    Storage Requirements Ambient temperature
    Connector Types BNC
    Description Sensing tip shape: Cylindrical
    For Use With PY-E50-2S; PY-B1L-2S; PY-A01-2S
    Filling Solution Type 3M KCl Potassium Chloride
    Warranty 1 Year
    Type pH combination microelectrode
    Reference Type Ag/AgCl
    Junction Platinum
    Material Glass

    Electrochemical analysis is one of the most frequent and demanding tasks in daily lab routines. Measurements will differ based on the composition, temperature, conductivity and viscosity of samples and it is these different conditions that set our electrodes apart from the rest. Whether for ultrapure water, jam, wine, TRIS Buffers, or drinking water, Sartorius has the right superior and dependable electrode for your sample types and application workflows. PlatinumLINE is our most robust and diverse electrode family that offer a variety of electrode types to fit all your unique sample types with superior construction, dependable design and accurate performance.

    Patented junction set sensors apart from the rest.

    • Reduce and/or eliminate metal ion contamination and chelation.
    • Ultimately minimizes damage to sensors, elongating the life of the electrodes, saving time and money.
    • Increase productivity and mitigate faulty measurements.
    • The highest quality materials result in better precision and accuracy.
    • The low resistance type A glass membranes are a perfect example of where quality materials interact with samples with less interference, yielding both speed and accuracy.
    • Liquid electrolyte electrodes with unique platinum junction and refill port slider for easier refilling and less evaporation of the electrolyte.
    • Air flush cap system prevents clogs from forming.
    • Individual serial number on each electrode for clear identification.
    • Flexibility and convenience.
    • Electrodes are available with integrated temperature sensors (ATC) or a separate temperature probe.
    • TRIS Compatibility and Impervious models available for Biological applications.
    PY-P20-2S
    Low Maintenance - pH, ceramic junction, gel filled

    PY-P21-2S
    High Performance - pH, Glass Body, Liquid 3M KCl refillable, air-flush platinum junction, TRIS compatible,break resistant conical shaped tip, ideal for biological applications, easy to clean

    PY-P22-2S
    Micro - 5mm diameter - pH, Liquid 3M KCl refillable, air flush platinum junction, TRIS,compatible ideal for low volume samples

    PY-P23-2S
    Flat-Surface - pH, Gel Filled, circular junction, ideal for meats, cheeses, agar, skin

    PY-P24-2S
    Sleeve - pH, Liquid 3M KCl refillable, Sleeve junction, ideal for low ionic samples

    PY-P27-2S
    Speartip - pH, hole junction, polymer filled

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit