Learn More
Sartorius™ pH Basic Benchtop Meters
Compact, basic equipment for routine measurements in the lab and reliable results in all applications
Manufacturer:  Sartorius™ PB-11-P10.1
 View more versions of this product
Catalog No. 14-557-203
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
PY-P10 Electrode
0.01
±1800mV
±0.02 to 0.05%mV
0.1°C
Integrated
Yes
32° to 212°F
pH/mV Meter
2 Years
0 to 14
±0.005
0.1mV
3
±0.2°C
BNC
-2 to +20pH
0° to 100°C
Plastic