Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius™ PlatinumLINE Electrode: pH

Fast and accurate results with high quality materials

Manufacturer:  Sartorius™ PYP272S/DEL

Catalog No. 14-557-896

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

  • Speartip
  • Plastic body
  • Hole junction
  • Poylmer gel filled
  • BNC connection
  • Cable: 1.5mm
Applications:
Cosmetics, Agriculture, Dairy, Food and beverage, Surface (skin and textiles), Drinking water, Detergents agents, Pharmaceutical/Biotech/Clinical/Pharmacy, Biology/Microbiology and Chemistry Lab, Extreme and difficult samples.
Specifications

Specifications

pH Electrode, Speartip
Polymer gel filled
1.5mm
PlatinumLINE
BNC
Hole Junction
Plastic
Documents
Promotions

Promotions

  • Sartorius , Fisher Scientific , Fisherbrand
    Buy $1,500 or more in Sartorius products and get a Sartorius Tacta Pipette Multipack at no additional cost.

    06-30-2021

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit