Learn More
Sartorius™ Polyethersulfone Membrane Filters Available on GSA/VA Contract for Federal Government customers only.
Polyethersulfone (PES) filters have a high internal porosity.
Manufacturer:  Sartorius™ 1540625N
 View more versions of this product
Catalog No. 14-555-454
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
0.5μm
25mm
Recommended for filtering biological and pharmaceutical solutions.
Polyethersulfone (PES)
100/pk.
150μm