Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius™ Premium Pro15 Benchtop pH Meters

Exact measurements with wider measuring ranges and better accuracy specifications

Manufacturer:  Sartorius™ PR15PHKIT/DEL

 View more versions of this product

Catalog No. 14-557-882

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

This advanced meter with German technology provides accurate measurements, wider ranges, autoCal and 5-point calibrations.
  • Large LCD display (B&W graphic w/lighting) with intuitive menu navigation
  • Automated calibration (Auto Cal) with up to 5-points
  • Automatic buffer recognition out of 22 pre-programmed buffer sets
  • Advance GLP Documentation with time and date stamps, administrator rights or user identification via an electronic ID card
  • ElectCheck (CalClock) and Stability Control indicates the condition of sensor, integrity of calibration and stability of reading at a single glance
  • User name can be assigned to each measured value
  • Wireless sensor recognition and user identification
  • Direct data transmission with RS232 and USB interfaces
  • Automatic switch-over to manual temperature input when no temperature sensor is connected
  • IQ/OQ documents available
Specifications:
  • Lower Housing: metal
  • Measuring Value Storage (manual/automatic): 1500 data sets, store intervals from 1 sec. to 60 min.
  • Plastic Foil Keypad with Tactile Response
  • Climate Class: 2 (VDI/VDE 3540)
  • Contrast Adjustment via Menu
  • Built-in real-time clock (processor solution), battery buffered, exchangeable battery
  • Power Supply: external universal power supply (medical approval) with country specific primary adapters (primary: 100 to 240V, 50/60Hz; secondary: 9V = 1.5A)
  • Display Size: 12 x 9cm
  • Dimensions (D x W x H): 24 x 28 x 8cm
  • Weight: approx. 2.5kg
pH Measurements:
  • Range: -2.0 to +20.0, -2.00 to +20.00, -2.000 to +20.000
  • Readability: 0.1, 0.01, 0.001
  • Accuracy: ±0.1, ±0.01, ±0.003
  • Dead Stop: Yes
  • Channels: One
  • Connector Type: BNC
  • Manually Enter Buffer: Yes
  • Reference Connector Type: 4mm banana plug
mV Measurements:
  • Range: -1999.9 to +1999.9, -2000 to +2000
  • Readability: 0.1, 1
  • Accuracy: ±0.2, ±1
  • Auto Range: Yes
  • Channels: One
Temperature Measurements:
  • Range: -10° to +120°
  • Readability: 0.1
  • Accuracy: ±0.1°C
  • Channels: One
  • Selectable °C/°F
  • Manually Adjustable: up to 130°C
  • Connector Types: 2.5mm phone jack connector for 10KOhm
ISE Measurements:
  • Range: 0.000 to 999,999mg/L
  • Readability: 0.001
  • Calibration Points: 2 to 5
  • Standard Concentration: 0.000 to 500,000mg/L
Includes:
  • Cat. No. 14-557-881 Pro15 Meter Kit: Meter, power supply, cover, arm, three buffers
  • Cat. No. 14-557-882 Pro15 pH Kit: Meter, power supply, cover, arm, three buffers, manual, PY-P11 electrode
  • Cat. No. 14-557-883 Pro15 Bio Kit: Meter, B&W backlit display, power supply, cover, arm, three buffers, manual, PY-P28 electrode

Certifications

CE marked

Compliance

cETLus, Protection class III, EG guidelines 73 / 23,EN 61010-1: 2001

Warranty and Services

Two years.

Specifications

Specifications

1500 Data Sets
Automatic/Manual
-10° to 120°C
±0.1, ±0.01, ±0.003
±0.2, ±1
Direct data transmission with RS232 and USB interfaces
24cm
External universal power supply (medical approval) with country specific primary adapters (primary: 120/240V, 50/60Hz; secondary: 9V = 1.5A)
-2.0 to 20.0, -2.00 to 20.00, -2.000 to 19.999
Metal
-1999.9 to 1999.9 mV, -2000 to 2000 mV
LCD
0.1
approx.2.5kg
28cm
±0.1°C
CE
8cm
0.1, 0.01, 0.001
Meter, power supply, cover, arm, three buffers, manual, PY-P11 electrode
0.1, 1
Documents
Promotions

Promotions

  • Sartorius , Fisher Scientific , Fisherbrand
    Buy $1,500 or more in Sartorius products and get a Sartorius Tacta Pipette Multipack at no additional cost.

    06-30-2021

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit