Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius™ Premium Pro50 Benchtop Multiparameter Meters

Exact measurements with wider measuring ranges and better accuracy specifications

Manufacturer:  Sartorius™ PR50A/DEL

 View more versions of this product

Catalog No. 14-557-886

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Includes:
  • Cat. No. 14-557-886 Pro50 Meter Kit: Meter with ID sensor Card, power supply, cover, arm and three buffers Cat. No. 14-557-887 Pro50 Bio Kit: Meter with ID sensor Card, power supply, cover, arm, three buffers, PY-P28 electrode

Description

Description

This advanced meter with German technology provides accurate measurements, wider ranges, autoCal and 5-point calibrations.
  • pH, ISE, DO, Conductivity and up to three parameters displayed simultaneously
  • Large LCD display (B&W graphic w/lighting) with intuitive menu navigation
  • Automated calibration (Auto Cal) with up to 5-points
  • Automatic buffer recognition out of 22 pre-programmed buffer sets
  • Advance GLP Documentation with time and date stamps, administrator rights or user identification via an electronic ID card
  • ElectCheck (CalClock) and Stability Control indicates the condition of sensor, integrity of calibration and stability of reading at a single glance
  • User name can be assigned to each measured value
  • Wireless sensor recognition and user identification
  • Direct data transmission with RS232 and USB (slave) interfaces
  • Automatic switch-over to manual temperature input when no temperature sensor is connected
  • IQ/OQ documents available


Specifications:
  • Lower Housing: metal
  • Plastic Foil Keypad with Tactile Response
  • Climate Class: 2 (VDI/VDE 3540)
  • Contrast Adjustment via Menu
  • Built-in real-time clock (processor solution), battery buffered, exchangeable battery
  • Power Supply: external universal power supply (medical approval) with country specific primary adapters (primary: 100 to 240V, 50/60Hz; secondary: 9V= 1.5A)
  • Battery Type: automatic switch-off 4 mignon battery (adjustable 10 min. to 24 hr., default 1 hr., cannot be switched off)
  • Display Size: 12 x 9cm
  • Dimensions (D x W x H): 24 x 28 x 8cm
  • Weight: approx.2.5kg
pH Measurements:
  • Range: -2.0 to +20.0, -2.00 to +20.00, -2.000 to +20.000
  • Readability: 0.1, 0.01, 0.001
  • Accuracy: ±0.1, ±0.01, ±0.003
  • Channels: One
  • Connector Type: BNC
  • Manually Enter Buffer: Yes
  • Reference Connector Type: 4mm banana plug


mV Measurements:
  • Range: -1999.9 to +1999.9, -2000 to +2000
  • Readability: 0.1, 1
  • Accuracy: ±0.2, ±1
  • Auto Range: Yes
  • Channels: One


Temperature Measurements:
  • Range: -10° to +120°
  • Readability: 0.1
  • Accuracy: ±0.1°C
  • Channels: One
  • Selectable °C/°F: Yes
  • Manually Adjustable: up to 130°C
  • Connector Types: 2.5mm phone jack connector for 10KOhm
ISE Measurements:
  • Range: 0.000 to 999,999mg/L
  • Readability: 0.001
  • Calibration Points: 2 to 5
  • Standard Concentration: 0.000 to 500,000mg/L


Conductivity Measurements:
  • Conductivity Range: 0.000µS/cm to 1000mS/cm
  • Salinity Range: 0.0 to 70.0 (acc. to IOT); 0.00 to 20 MOhm cm
  • TDS Range: 0 to 1999mg/L
  • Readability: 0.01
  • Accuracy: ±0.5 % of value (dependent upon probe)
  • TDS factor 0.4 to 1.0
  • Salinity UNESCO1966n Natural Sea Water Scale
  • Accuracy in % from Measuring Value (±1 dig): 0.5
  • Calibrated cell constant 0.450 to 0.500cm-1; 0.585 to 0.715cm-1; 0.800 to 1.200cm-1(calibration with control standard) d 0.01 mol KCl
  • Adjustable cell constant 0.250 to 2.500cm-1 and 0.090 to 0.010cm-1
  • Fixed Cell Constant 0.010cm-1
  • Temperature Compensation in Pure Water
  • Reference Temp. 20° or 25°C Selectable


DO Measurements:
  • O2 Concentration Range: 0 to 20.00mg/L, 0 to 90.0mg/L
  • O2 Concentration Accuracy: 0.01mg/L, 0.1mg/L
  • O2 Saturation Range: 0 to 200.0%, 0 to 600%
  • O2 Saturation Accuracy: 0.1%, 1%
  • O2 Partial Pressure Range: 0 to 200.0mbar, 0 to 1250mbar
  • O2 Partial Pressure Accuracy: 0.1mbar, 1mbar
  • Accuracy in % from measuring value (±1 digit) at ambient 5° to 30°C: 0.5
  • Salinity Correction
  • Calibration in calibration vessel with water vapor-saturated air

Certifications

CE marked

Compliance

cETLus, Protection class III, EG guidelines 73/23, EN 61010-1: 2001

Warranty and Services

Two years.

Specifications

Specifications

Meter with ID sensor Card, power supply, cover, arm and three buffers
0.1, 0.01, 0.001
-1999.9 to +1999.9, -2000 to +2000
±0.2, ±1
0.001
0 to 1999mg/L
5-Points
-10° to +120°
±0.1°C
RS232 and USB (slave) interfaces
approx.2.5kg
8cm
Pro50
approx. 3kg
28cm
-2.0 to +20.0, -2.00 to +20.00, -2.000 to +20.000
±0.1, ±0.01, ±0.003
0.1, 1
0.000 to 999,999mg/L
0.000μS/cm to 1000mS/cm
0.0 to 70.0 (acc. to IOT); 0.00 to 20 MOhm cm
2 to 5
0.1
LCD
CE marked
24cm
50/60Hz
0 to 20.00mg/L, 0 to 90.0mg/L
100 to 240V
Documents
Promotions

Promotions

  • Sartorius , Fisherbrand , Fisher Scientific
    Buy $1,500 or more in Sartorius products and get a Sartorius Tacta Pipette Multipack at no additional cost.

    06-30-2021

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit