Sartorius™ Professional Benchtop Meters

For professional measurements and optimal testing in laboratories

Manufacturer: Sartorius™ PP20

 View all options for this product

Catalog No. 14-557-200


This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Range (pH) -2 to 20
    Resolution (pH) ±0.001
    Accuracy (pH) ±0.001
    Range (mV) ±2000mV
    Resolution (mV) 0.1/2mV
    Accuracy (mV) ±0.05mV
    Range (ISE) 1 to 9 x 10 to 9ppm
    Range (Conductivity) 0 to 300000μS/cm
    Range (TDS) 0.005 to 300000
    Range (Salinity) 0.01 to 42ppt
    Data Points 620
    Inputs BNC, DIN
    Depth (Metric) 20cm
    Detection Range -2 to +20pH
    Height (English) 3.93 in.
    Height (Metric) 10cm
    Width (Metric) 26.5cm
    Width (English) 10.43 in.

    Easy to operate with the highest measurement accuracy and data management.

    • Complete GLP/GMP Documentation
    • Automatic buffer recognition
    • Automatic temperature compensation
    • Automatic electrode function check
    • Calibration reminder
    • Data memory for up to 620 data records
    • Bidirectional interface (RS232) to computer, computer, LIMS, autosampler etc.

    pH Range: 0-14pH
    pH Resolution: 0.001/0.01/0.1
    pH Accuracy: +/-0.001pH

    mV Range:+/-2400mV
    mV Resolution: 0.1/2
    mV Accuracy: +/-0.05

    Ion Range: 1 to 9x10-9
    Ion Resolution: 1,2,3 signif. fig.
    Ion Accuracy: +/-0.9n%

    Conductivity Range:0 to 300,000 S/µcm
    ConductivityIon Resolution:1,2,3 signif. fig.
    Conductivity Accuracy: +/-0.5% of reading

    Temp.: -5.0 to 105°C

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit