Learn More
Sartorius™ PTFE (Polytetrafluorethylene) Membrane Filters Available on GSA/VA Contract for Federal Government customers only.
Filter air and gasses, even at low differential pressures. Sartorius™ PTFE (Polytetrafluorethylene) Membrane Filters are permanently hydrophobic with low adsorption and excellent chemical compatibility. Choose from four pore sizes in a variety of diameters.
Manufacturer:  Sartorius™ 1180313N
 View more versions of this product
Catalog No. 14-555-410
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
1.2μm
13mm
100/pk.
0.45bar
80mL/min. (Isopropanol)
Hydrophobic
Polytetrafluoroethylene (PTFE)
White
Yes
Air/gas-filtration
100µm