Learn More
Sartorius™ Quantitative Grade 1288 Filter Papers
Manufacturer:  Sartorius™ FT3206500
 View more versions of this product
Catalog No. 14-555-073
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Made of refined pulp and cotton linters with an > 95% α-cellulose content. Ash content < 0,1% according to DIN 54370. Wet-strengthened.
Specifications
Specifications
100 per pack
Disc
12 to 15μmm
1288
500mm
0.21mm