Learn More
Sartorius™ Quantitative Grade 1289 Filter Papers
Manufacturer:  Sartorius™ FT3207185
 View more versions of this product
Catalog No. 14-555-065
Add to cart
Description
Description
Made of refined pulp and cotton linters with an > 95% α-cellulose content. Ash content < 0,1% according to DIN 54370. Wet-strengthened.
Specifications
Specifications
100 per pack
Disc
8 to 12μmm
1289
185mm
0.21mm