Learn More
Sartorius™ Quantitative Grade 292 Filter Papers
Manufacturer:  Sartorius™ FT3205500
 View more versions of this product
Catalog No. 14-555-084
Add to cart
Description
Description
  • Pure cotton linters or cotton linters with refined pulp
  • No additives, such as wet-strengthening agents
Specifications
Specifications
100 per pack
Disc
5 to 8μmm
292
500mm
0.18mm