Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius™ Quantitative Grade 391 Filter Papers

Used for quantitative and gravimetric analyses, as well as for pressure or vacuum filtration. Sartorius™ Qualitative Grade 391 Filter Papers are made of 100% cotton linters with an α-cellulose content of >98%.

Manufacturer:  Sartorius™ FT3104125

 View more versions of this product

Catalog No. 14-555-208


Add to cart

Description

Description

  • Acid-washed to achieve high purity
  • Ash-free (ash content ≤ 0.01 % according to DIN 540370)
  • Wet-strengthened
  • Color-coded box for easy selection
Specifications

Specifications

100 per pack
Cotton linters
125mm
quantitative and gravimetric analyses, pressure or vacuum filtration
0.15mm
391
Filter Discs
2 to 3μmm
Blue
84g/m2
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit