Learn More
Sartorius™ Quantitative Grade 393 Filter Papers
Manufacturer:  Sartorius™ FT3127185
 View more versions of this product
Catalog No. 14-555-107
Add to cart
Description
Description
Made of 100% cotton linters. Ash-free (ash content ≤ 0.01 % according to DIN 540370). Wet-strengthened. Color-coded box for easy selection.
Specifications
Specifications
100 per pack
Disc
1 to 2μmm
0.18mm
393
185mm
Purple