Learn More
Sartorius™ Regenerated cellulose (RC) Membrane Filters Available on GSA/VA Contract for Federal Government customers only.
These solvent-resistant, hydrophilic membrane filters are excellently suited for particle removal from solvents.
Manufacturer:  Sartorius™ 1840650N
 View more versions of this product
Catalog No. 14-555-466
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
0.5μm
50mm
Yes
160 to 200μm
Regenerated Cellulose (RC)
100/pk.
For particle removal from solvents.
Hydrophilic