Learn More
Sartorius Regenerated cellulose (RC) Membrane Filters
These solvent-resistant, hydrophilic membrane filters are excellently suited for particle removal from solvents.
Supplier:  Sartorius 1840725N
 View more versions of this product
Catalog No. 14-555-468
Add to Cart
Description
Description
These solvent-resistant, hydrophilic membrane filters are excellently suited for particle removal from solvents.
Regenerated cellulose membrane filters are reinforced with nonwoven cellulose. The membranes are mainly used for particle removal from solvents. For example, the 50mm diameter, 0.45μm pore size filter is standardly used to ultraclean and degas solvents and mobile phases for HPLC, in combination with an all-glass filter holder. Regenerated cellulose membrane also feature low non-specific-adsorption.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Autoclavable
Regenerated Cellulose
0.2 μm
Hydrophilic
160 to 200 μm
25 mm
Membrane Filter
Cellulose
For particle removal from solvents.
100/Pk.
Membrane Filter
Product Suggestions
Product Suggestions