Learn More
Sartorius™ Multi-Range Conductivity/ATC Electrode with RFID
Multi range, target configuration Conductivity/ATC electrode.
Manufacturer:  Sartorius™ PYC132S
Catalog No. 14-557-900
Add to cart
Description
Description
Conductivity measuring cells are used for the conductivity measurement of fluids, such as pure and ultra pure water. Epoxy Bodies and graphite electrodes feature the highest grade of robustness and long-term stability and offer the widest measuring range. The conductivity measuring cells are suited for temperature measurement and are equipped with automatic temperature compensation.
Cells offer a high degree of precision, diversity in wider measuring ranges. Cell geometry design address and minimize polarization effects with difficult sample types.
Note: Only compatible with Sartorius PR-20 and PR-50 meters
  • Optimized cell geometry with 2, 4 band and target configurations
  • Diverse and wider measuring ranges
  • Long term cell constant stability
  • Robust and durable due to high quality epoxy and graphite construction allowing for break and abrasion resistant body materials
  • Integrated temperature compensation
  • Smart design mitigates measuring error from cable interferences, polarizations effects, air bubble and vessel influences
Specifications
Specifications
1 year
Epoxy Body, Graphite electrode, 4 graphite rings
Target 0.475 with ID
Integrated ATC NTC 30 K Ohm
Range: 1 to 2mS/cm
Sartorius PR-20 and PR-50 meters
ambient - room temp
Documents
Promotions
Promotions
  • Sartorius , Fisher Scientific , Fisherbrand
    Buy $1,500 or more in Sartorius products and get a Sartorius Tacta Pipette Multipack at no additional cost.
    06-30-2021