Learn More
Sartorius™ Sartolab™ 150v Vacuum Filtration Units
Optimised for the application in cell culture
Manufacturer:  Sartorius™ 18080M
Catalog No. 14-555-992
Add to cart
Description
Description
Sartolab 150v is a disposable, 0.2µm sterile, ready-to-use membrane filter capsule for highest convenience. Sartolab 150v capsules are made of a unique hydrophilic Polyethersulfone (PES) membrane providing outstanding total throughput, flow rate performance, low extractables and broadest chemical compatibility. Each 0.2 ++m bottle-top capsule filters 15 liters of aqueous solutions. The integrated on-off valve switch allows easy movement between receiving flasks.
  • Quality and reliability
  • Low protein binding
  • Fast flow and high throughput
  • Easy to use
Specifications
Specifications
Not Autoclavable
Flask Rack
Documents
Promotions
Promotions
  • Sartorius , Fisher Scientific , Fisherbrand
    Buy $1,500 or more in Sartorius products and get a Sartorius Tacta Pipette Multipack at no additional cost.
    06-30-2021