Sartorius™ Sartolab™ RF/BT Vacuum Filtration Units Available on GSA/VA Contract for Federal Government customers only.

Filter aqueous solutions quickly and easily. Sartorius™ Sartolab™ RF/BT Vacuum Filtration Units are disposable and easy to use filter units. They contain a high quality PES membrane and combine the highest flow-rates and throughput with extremely low protein binding and extractables. Available in a range of volumes.

Manufacturer: Sartorius™ 180C2E

 View more versions of this product

Catalog No. 14-555-994

  Check Availability
Add to Cart

    Specifications

    Quantity (No. per Case) 12 Case
    Product Type Sartolab RF Filter
    Pore Size 0.22μm
    Sterile Yes
    Certifications/Compliance USP Class VI, ISO 10993
    Connectors Plug-on hose nipple
    Disposable Yes
    Filter Material Polyethersulfone
    Filtration Area 39cm2
    For Use With (Application) Filtration of tissue culture media, buffers, additives and other aqueous biologicals
    For Use With (Equipment) 1.77 in. bottle neck
    Material Polystyrene
    Volume (Metric) 500mL

    Quality, reliable sterile filtration units

    • Receiver Flask (RF) and Bottle Top (BT) units contain a high quality Sartorius PES membrane
    • Combine the highest flow-rates and throughput with extremely low protein binding and extractables
    • Ideal for filtration of tissue culture media, buffers, additives and other aqueous biologicals
    • RF and BT vacuum units are available as 150mL, 250mL, 500mL and 1000mL units
      • 500mL and 1000mL units come standard with 90mm membranes
    • Improved filtration speed and throughput for cost effective filtration

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit