Learn More
Sartorius™ Sartolab™ RF/BT Vacuum Filtration Units Available on GSA/VA Contract for Federal Government customers only.
Filter aqueous solutions quickly and easily. Sartorius™ Sartolab™ RF/BT Vacuum Filtration Units are disposable and easy to use filter units. They contain a high quality PES membrane and combine the highest flow-rates and throughput with extremely low protein binding and extractables. Available in a range of volumes.
Manufacturer:  Sartorius™ 180C3E
 View more versions of this product
Catalog No. 14-555-995
  Check Availability
Add to cart
Specifications
Description
Quality, reliable sterile filtration units
  • Receiver Flask (RF) and Bottle Top (BT) units contain a high quality Sartorius PES membrane
  • Combine the highest flow-rates and throughput with extremely low protein binding and extractables
  • Ideal for filtration of tissue culture media, buffers, additives and other aqueous biologicals
  • RF and BT vacuum units are available as 150mL, 250mL, 500mL and 1000mL units
    • 500mL and 1000mL units come standard with 90mm membranes
  • Improved filtration speed and throughput for cost effective filtration
Description & Specifications
Specifications
USP Class VI, ISO 10993
Yes
62cm2
1.77 in. bottle neck
Sartolab RF Filter
0.22μm
Yes
Plug-on hose nipple
Polyethersulfone
Filtration of tissue culture media, buffers, additives and other aqueous biologicals
Polystyrene
1000mL
12 Case