Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sartorius Sartolab™ RF/BT Vacuum Filtration Units
Filter aqueous solutions quickly and easily. Sartorius Sartolab™ RF/BT Vacuum Filtration Units are disposable and easy to use filter units. They contain a high quality PES membrane and combine the highest flow-rates and throughput with extremely low protein binding and extractables. Available in a range of volumes.
Specifications
Certifications/Compliance | USP Class VI, ISO 10993 |
---|---|
Connectors | Plug-on hose nipple |
Disposable | Single-use |
For Use With (Equipment) | 1.77 in. bottle neck |
Material | Polystyrene |
Catalog Number | Mfr. No. | Type | Filtration Area | Volume (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Filtration Area | Volume (Metric) | Price | Quantity & Availability | |||
14-555-995
|
Sartorius
180C3E |
Sartolab RF Filter | 62 cm2 | 1000 mL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
Description
Quality, reliable sterile filtration units- Receiver Flask (RF) and Bottle Top (BT) units contain a high quality Sartorius PES membrane
- Combine the highest flow-rates and throughput with extremely low protein binding and extractables
- Ideal for filtration of tissue culture media, buffers, additives and other aqueous biologicals
- RF and BT vacuum units are available as 150mL, 250mL, 500mL and 1000mL units
- 500mL and 1000mL units come standard with 90mm membranes
- Improved filtration speed and throughput for cost effective filtration
Specifications
USP Class VI, ISO 10993 | |
Single-use | |
Polystyrene | |
0.22 μm | |
Filtration of tissue culture media, buffers, additives and other aqueous biologicals |
Plug-on hose nipple | |
1.77 in. bottle neck | |
PES | |
Sterile |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title