Learn More
Sartorius™ Stedim Phase Separating Filter Paper
Grade 480 is impregnated with stabilized silicon, thus rendering it hydrophobic: It retains water, but allows solvents to flow through. The flow stops automatically when the entire solvent has passed through. In many applications, this phase separator paper eliminates the need to use separating funnels.
Manufacturer:  Sartorius™ FT3602150
Catalog No. 14-555-016
Add to cart
Description
Description
Applications include: Filtration of organic solvents that have been contaminated with water to separate the aqueous from the organic phase, Filtration of extracting solvents in clinical or medical labs, Separation of emulsions that are formed during the extraction of aqueous plant or drug solutions.
Specifications
Specifications
150mm
Separates aqueous from organic phases
0.19mm
480
Disc
Documents
Promotions
Promotions
  • Sartorius , Fisherbrand , Fisher Scientific
    Buy $1,500 or more in Sartorius products and get a Sartorius Tacta Pipette Multipack at no additional cost.
    06-30-2021