Learn More
Sartorius™ Traditional Filter Holders and Manifolds Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  Sartorius™ 16824
 View more versions of this product
Catalog No. 14-555-791
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
3.38 oz.
ISO 8199, B.S. 304S31, AISI 304
12cm
4.7/5cm
7.24 in.
Stainless Steel funnels and lids
2 bar
3 x 10mL
43.2cm
3 Branch Manifold
100mL
4.7 in.
1.85/1.96 in.
12.5cm2
18.4cm
Stainless Steel
Hose Nipple, DN 10
3 or 6
Specifically for applications in which the particles or microorganisms retained on the membrane filter surface are of interest