Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sartorius Vivaspin™ 100 Disposable Ultrafiltration Units
Click to view available options
MWCO:
0.2 μm
10 kDa
100 kDa
30 kDa
300 kDa
5 kDa
50 kDa
Quantity:
10 Pk.
2 Pk.
Description
- For the concentration of samples up to 98 mL
- Can be used either in a centrifuge or for pressure filtration
- Combines the ease of use of centrifugal devices with the flexibility and control provided by pressurized ultrafiltration cells
- Easy to assemble
- Requires no tubing connections or stirring mechanisms
- Can be adapted to equipment availability or to specific user preferences
- For convenience, the units are simply spun in a large capacity centrifuge (carriers accepting 250 mL bottles)
- For better process control, gas pressure can be combined with gentle orbital shaking
- For highest simplicity of use with minimum equipment requirements, the device is pressurized and then left standing on a bench top or in a refrigerator
- The concentrator features a vertical membrane that minimizes fouling, while the built-in deadstop of 350 μL will prevent concentration to dryness.
Features:
- Easy to handle
- High flux PES membrane
- Vertical twin membrane for ultra-fast concentration
- High retentate recovery (90%)
- Total process flexibility
- Compatible with swing out rotors

Specifications
Specifications
Disposable | Single-use |
Filter Type | Ultrafiltration |
For Use With (Equipment) | Centrifuge |
Material (Membrane) | PES |
MWCO | 10 kDa |
Sample Volume | 100 mL |
Sterility | Non-sterile |
Max. Force | 5 bar |
Quantity | 2 Pk. |
Type | Centrifugal Concentrator |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction