Learn More
Sartorius™ Vivacon™ 2 DNA Concentrator Available on GSA/VA Contract for Federal Government customers only.
Centrifugal units for fast DNA concentration and buffer exchange of 2 ml samples
Manufacturer:  Sartorius™ VN02H03
 View more versions of this product
Catalog No. 14-558-362
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
60 min.
4.92 x 0.62 in. (12.5 x 1.6cm)
4.52 in.
11.5cm
Polycarbonate Body, Polypropylene filtrate vessel, Polypropylene back spin vial, Polypropylene concentrator cap, Hydrosart™ membrane, Silicone O-ring
10000 MWCO
DNA, peptide
0.62 in.
DNA concentration, desalting, buffer exchange, fractionation
500 Pack
50ng/mL
0.95cm2
4.92 in.
12.5cm
5000xG
30 bp
10μL
1.6cm
Vivacon™ 2 DNA Concentrator
2mL