Sartorius™ Vivacon™ 2 DNA Concentrator Available on GSA/VA Contract for Federal Government customers only.

Centrifugal units for fast DNA concentration and buffer exchange of 2 ml samples

Manufacturer: Sartorius™ VN02H33

 View more versions of this product

Catalog No. 14-558-371

  Check Availability
Add to Cart

    Specifications

    Quantity 500 Pack
    Capacity (Metric) 2mL
    Concentration Time 45 min.
    Concentration 50ng/mL
    Dimensions (L x W) 4.92 x 0.62 in. (12.5 x 1.6cm)
    Filtration Area 0.95cm2
    Length (English) Back Spin 4.52 in.
    Length (English) 4.92 in.
    Length (Metric) Back Spin 11.5cm
    Length (Metric) 12.5cm
    Material Polycarbonate Body, Polypropylene filtrate vessel, Polypropylene back spin vial, Polypropylene concentrator cap, Hydrosart™ membrane, Silicone O-ring
    Max. Force 2500xG
    Molecular Weight 50000 MWCO
    Sample Size 300 bp
    Sample Type DNA, peptide
    Volume (Metric) Hold Up 10μL
    Width (English) 0.62 in.
    Width (Metric) 1.6cm
    For Use With (Application) DNA concentration, desalting, buffer exchange, fractionation
    Product Type Vivacon™ 2 DNA Concentrator

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit