Sartorius™ Vivacon™ 500 DNA Concentrator Available on GSA/VA Contract for Federal Government customers only.

Manufacturer: Sartorius™ VN01H43

 View more versions of this product

Catalog No. 14-558-357

  Check Availability
Add to Cart

    Specifications

    Dimensions (L x W) 1.77 x 0.48 in. (4.5 x 1.24cm)
    Max. Force 3000xG
    Membrane Material Hydrosart Regenerated Cellulose
    Molecular Weight 100000 MWCO
    Quantity 500 Pack
    Capacity (Metric) 0.5mL
    Concentration Time 10 min.
    Concentration 50ng/mL
    Filtration Area 0.32cm2
    Length (English) Back Spin 1.87 in.
    Length (English) 1.77 in.
    Length (Metric) Back Spin 4.75cm
    Length (Metric) 4.5cm
    Material Polycarbonate Body, Polypropylene filtrate vessel, Hydrosart™ membrane
    Sample Size 600 bp
    Sample Type DNA, peptide
    Volume (Metric) Hold Up <5μL
    Width (English) 0.48 in.
    Width (Metric) 1.24cm
    For Use With (Application) DNA concentration, desalting, buffer exchange, fractionation
    Product Type Vivacon™ 500 Concentrator

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit