Learn More
Sartorius Vivapore™ 10/20 Static Concentrators
Ideal for general purpose laboratory concentration or purification prior to further analysis.
$59.00 - $1071.00
Specifications
Material (Membrane) | PES |
---|---|
MWCO | 7.5 kDa |
Filtration Area | 28 cm2 |
Volume (Metric) | 2 to 10 mL, 20 mL |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
14-558-390
|
Sartorius
VP2003 |
4/Pk. |
Pack of 4 for $59.00
|
|
|||||
14-558-388
|
Sartorius
VP2001 |
30/Pk. |
Pack of 30 for $413.00
|
|
|||||
14-558-389
|
Sartorius
VP2002 |
100/Pk. |
Pack of 100 for $1,071.00
|
|
|||||
Description
Vivapore 10/20 is a ready-to-use benchtop concentrator for samples up to 20mL. With no need for additional equipment, pressure or vacuum source, solvent absorption is the most economic and user friendly concentration technique available for clinical and general research applications. Just fill the unit with the solution to be concentrated, wait for the desired concentration level to be achieved and then pipette the concentrated sample from the dead-stop reservoir at the bottom of the concentrator. Concentration by Vivapore is particularly suited for samples that are sensitive to shearing forces or to pressure-induced stress or that require processing in a cold room environment. Vivapore static concentrators are offered as separate chambers together with disposable plastic stands for up to four units. Contrary to other, larger products with multiple, adjacent concentration channels, there is no risk of cross-contamination between individual Vivapore units. Vivapore 10/20 is frequently used for urine concentration and subsequent analysis of Bence-Jones proteins by gel electrophoresis. Each unit has a 7,500 MWCO PES membrane.
- Sample concentration without additional equipment
- High-flow membrane
- Seperate chambers/no cross contamination
- Very gentle concentration process
- Up to 20mL sample volume
- Desalting/diafiltration option
Specifications
PES | |
7.5 kDa | |
28 cm2 | |
2 to 10 mL, 20 mL |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.