Sartorius™ Vivapore 5 Static Concentrators Available on GSA/VA Contract for Federal Government customers only.

Designed for quick and affordable concentration. Sartorius™ Vivapore 5 Static Concentrators are ready to use, disposable ultrafiltration devices for samples up to 5mL. These simple benchtop units are ideal for general purpose laboratory concentration or purification prior to further analysis. Available in packages of 4, 30 or 100.

Manufacturer: Sartorius™ VP0503

 View more versions of this product

Catalog No. 14-558-387

  Check Availability
Add to Cart

    Specifications

    Housing Material Styrene acrylonitrile (SAN)
    Length (Metric) 82mm
    Disposable Yes
    Width (Metric) 42mm
    Membrane Material Polyethersulfone (PES)
    MWCO 7,500 d
    Quantity 4/Pk.
    Surface Area 20cm2
    Volume (Metric) 1 to 5mL

    Economic and user-friendly

    • No need for additional equipment, pressure or a vacuum source
    • Individual devices with disposable plastic stands for up to four units
    • Simple concentration technique:
      • Just fill the unit with the solution to be concentrated
      • Wait for the desired concentration level to be achieved
      • Then pipet the concentrated sample from the dead-stop reservoir
    • Separate concentration chambers eliminate the risk of cross-contamination
    Gentle, reliable concentration process
    • Each unit features a high-flow, 7.5 kDa PES membrane
    • Ideal for samples that are sensitive to shearing forces or to pressure-induced stress
    • Well suited for samples that require processing in a cold room environment
    • Perfect for a variety of clinical and general research applications
      • Urine concentration and subsequent analysis of Bence-Jones proteins by gel electrophoresis
      • Concentrating macromolecules in cerebrospinal fluid (CSF) or other biological solutions

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit