Sartorius™ Vivapure™ maxiprep MC spin columns Available on GSA/VA Contract for Federal Government customers only.

Larger spin columns for simple, rapid purification of His-tagged recombinant proteins from cell lysates using Ni2+-IDA agarose resin for samples up to 20 ml and one re-use

Manufacturer: Sartorius™ VSMCMAXIK

 View more versions of this product

Catalog No. 14-558-568

  Check Availability
Add to Cart

    Specifications

    Product Type MaxiprepMC spin column Kit
    Quantity 8 Pack
    For Use With (Application) Rapid purification of His-tagged recombinant Proteins from cell lysates by Metal chelate affinity chromato graphy
    Includes UF-concentrato rs and Buffers
    Sufficient For 2 Users

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit