Learn More
Sartorius Vivapure™ Mini S&Q H Starter Kit
Ready-to-use centrifugal devices
Supplier:  Sartorius VSIX01SQ16
Catalog No. 14-558-557
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
Vivapure Mini Ion Exchange spin columns are centrifugal devices, based on Sartobind Membrane Adsorber technology as their chromatography matrix. The devices are ready-to-use and cannot run dry by their design. For many protein purification applications, they can replace time-consuming column chromatography. The rapid 1-2-3 bind-wash-elute protocol especially lends itself to screening applications, where many samples are processed parallel. Vivapure Mini is available with three different membrane chemistries and with medium (1mg/unit) or high (4mg/unit) protein binding capacity. Vivapure Mini units are intended for use in swing-out rotors and accommodate 0.5mL and 0.4mL samples, respectively.
  • Fast centrifugal spin columns
  • Easy and ready-to-use
  • Binding capacity 1mg up to 4mg protein
  • Ideal for parallel processing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
240 μL
Polypropylene
Stabilized regenerated Cellulose
Sample fractionation, purification condition scouting, small scale purification
0.4 mL
3 to 5 μm
16 Pack
Spin Column
Product Suggestions
Product Suggestions