Sartorius™ Vivapure™ miniprep A / G spin columns Available on GSA/VA Contract for Federal Government customers only.

Centrifugal units with Protein A or Protein G resin for fast affinity purification of antibodies from small samples up to 0.65 ml.

Manufacturer: Sartorius™ VSARGBUFPK

 View more versions of this product

Catalog No. 14-558-540

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) Purification of Monoclonal and Polyclonal Antibodies from Serum, ascites and cell culture supernatants
    Product Type Buffer Pack
    Quantity Each
    Sufficient For 4 Maxi or 48 miniprep Protein A and G spin column

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit