Sartorius™ Vivapure™ miniprep MC spin columns Available on GSA/VA Contract for Federal Government customers only.

Spin columns for simple, rapid purification of His-tagged recombinant proteins from cell lysates using Ni2+-IDA agarose resin for samples up to 0.65 ml and one re-use

Manufacturer: Sartorius™ VSMCMINIB

 View more versions of this product

Catalog No. 14-558-570

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) Rapid purification of His-tagged recombinant Proteins from cell lysates
    Product Type MiniprepMC spin column Kit
    Quantity 72 Pack
    Sufficient For 2 Users

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit