Sartorius™ Vivaspin™ 2 Centrifugal Concentrators: PES Membrane Available on GSA/VA Contract for Federal Government customers only.

Combines high speed with superior retentate recovery for concentrating very dilute solutions. Sartorius™ Vivaspin™ 2 Centrifugal Concentrators: PES Membrane feature twin vertical membranes, small internal surface areas and thin-channel chambers. Available in a range of MWCO and packaging quantities.

Manufacturer: Sartorius™ VS0242

 View more versions of this product

Catalog No. 14-558-426

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) Concentration, desalting and buffer applications
    Housing Material Polycarbonate
    Max. Force Swing bucket rotor: 4,000 x g
    Fixed angle rotor: 9,000 x g (3 - 50kDa MWCO); 12,000 x g (100kDa+ MWCO)
    Surface Area 1.3cm2
    Width (Metric) 17.8mm
    Length (Metric) 126mm
    MWCO 100,000 d
    Quantity 100/Pk.
    Disposable Yes
    Volume (Metric) 2mL (Fixed Angle Rotor); 3mL (Swing Bucket Rotor)
    Membrane Material Polyethersulfone

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit