SAS™ Legionella Test

Aids in the detection of Legionnaire's disease

Manufacturer: SA ScientificSupplier Diversity Partner 044020

 View more versions of this product

Catalog No. 22-100-002

  Check Availability
Add to Cart

Required Accessories : Timer; Specimen Collection Container

    Specifications

    Product Type Legionella Rapid Assay Kit
    CPT Code 87449
    Quantity 20 Tests
    Sensitivity 91%
    Specificity 0.95
    Storage Requirements Room temperature
    Clia Complexity Non-waived
    Detectable Analytes Legionella pneumophila serogroup 1 Antigens in Urine
    Includes Test device, disposable specimen droppers
    Test Time >15 min.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit