Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sash1 Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549173

Catalog No. PA549173

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

May have a role in a signaling pathway. Could act as a tumor suppressor.
Specifications

Specifications

Sash1
Polyclonal
PBS with 0.09% sodium azide
P59808
SAM and SH3 domain-containing protein 1, Sash1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
70097
Western Blot
Unconjugated
Sash1
Liquid
SASH1
KLH conjugated synthetic peptide between 870-896 amino acids from the C-terminal region of mouse Sash1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit