Learn More
Aviva Systems Biology Corp SASP Antibody - middle region (ARP53113_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP53113_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SASP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SASP. Alternate names: MUNO, SASPase, Taps, SASPApplicationWB. Species reactivity: Guinea Pig, Horse, Human, Rat
Catalog No. 50-014-60927