Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SAT2 Mouse anti-Human, Mouse, Clone: OTI2B3, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA504632

Catalog No. 50-167-5278


Add to cart

Description

Description

SAT2 is an enzyme which catalyzes the acetylation of polyamines. Substrate specificity: norspermidine > spermidine = spermine >> N(1)acetylspermine = putrescine..
Specifications

Specifications

SAT2
Monoclonal
0.58 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q6P8J2, Q96F10
SSAT2
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
112483, 69215
Immunohistochemistry (Paraffin), Western Blot
OTI2B3
Unconjugated
SAT2
Liquid
Sat2
Full length human recombit protein of human SAT2 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human, Mouse
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit