Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SAT2 Mouse anti-Human, Non-human primate, Clone: OTI2A3, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525939

Catalog No. PIMA525939


Add to cart

Description

Description

SAT2 is an enzyme which catalyzes the acetylation of polyamines. Substrate specificity: norspermidine > spermidine = spermine >> N(1)acetylspermine = putrescine.
Specifications

Specifications

SAT2
Monoclonal
4.45 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q96F10
Diamine acetyltransferase 2; Polyamine N-acetyltransferase 2; Spermidine/spermine N(1)-acetyltransferase 2; Thialysine N-epsilon-acetyltransferase; SAT2; SSAT2
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
112483, 716044
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI2A3
Unconjugated
SAT2
Liquid
SAT2
Full length human recombit protein of SAT2 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human, Non-human primate
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit