Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SATB2 Rabbit anti-Human, Clone: SATB2/4374R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen SATB2
Clone SATB2/4374R
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122448
View Documents
Novus Biologicals
NBP30748720UG
20 μg
Each for $159.82
Add to cart
 
NB122447
View Documents
Novus Biologicals
NBP30748710UG
100 μg
Each for $406.52
Add to cart
 
Description

Description

SATB2 Monoclonal antibody specifically detects SATB2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

SATB2
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
10 mM PBS with 0.05% BSA
23314
IgG
Primary
Store at 4°C.
SATB2/4374R
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Rabbit
Human
DNA-binding protein SATB2, FLJ32076, KIAA1034FLJ21474, MGC119474, MGC119477, SATB family member 2, SATB homeobox 2, Special AT-rich sequence-binding protein 2
Synthetic peptide corresponding to residues within aa200-300 corresponding to human SATB2
Monoclonal
Protein A or G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit