Learn More
Berkshire SatPax™ 550 Wipes
SatPax™ 550 wipers are pre-wetted with a consistent concentration of IPA and DI water, providing a cost-effective and easy-to-use solution versus traditional bulk handling of solvents, maintenance of squirt bottles and inconsistent wetting and cleaning associated with wetting a dry wiper.
Supplier: Berkshire SPX55000836
Description
- Very low extractables and ions
- Very low particle and fiber counts
- Excellent wet strength and abrasion resistance
- Reduces alcohol usage and preparation / handling costs
- Reduces VOC emissions
- Increases cleaning efficiency
- Increases cleaning protocol consistency
- Re-sealable solvent resistant packaging
- Wipers in c-folded configuration for single withdrawal
Specifications
Cleanroom Wiper | |
28 cm | |
18 cm | |
Not Autoclavable | |
Yes | |
Non-sterile |
11 in. | |
7 in. | |
Meltblown Polypropylene | |
ISO Class 5 - 8, Cleanroom Class 100 - 100,000, EU Grade B/C/D, USP <797> | |
1-ply |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.