Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Berkshire™ SatPax™ MicroPolx™ Nonwoven Cleanroom Wipes

Berkshire™ SatPax™ MicroPolx™ Nonwoven Cleanroom Wipes featuring a leading edge innovative manufacturing process of 100% polyester microfiber wet-laid nonwoven performance material.

$33.67 - $34.69

Specifications

Color White
Dimensions (L x W) 12 x 12 cm (9 x 9 in.)
For Use With (Application) ISO class 5 and above applications
Material Polyester Microfiber
Product Type Nonwoven Cleanroom Wipes
View More Specs

Products 2
Catalog Number Mfr. No. Saturation Solution Price Quantity & Availability  
Catalog Number Mfr. No. Saturation Solution Price Quantity & Availability  
19-066-745
View Documents
Berkshire™
SPXMPNW500112
45% Pack for $33.67
Case of 12 PK for $258.56
Add to cart
 
19-066-746
View Documents
Berkshire™
SPXMPNW500112R
60% Pack for $34.69
Case of 12 PK for $277.64
Add to cart
 
Description

Description

  • Extremely soft hand
  • Mechanical strength
  • Anti-scratch
  • Multi-directional layering
  • Chemical compatibility
  • Lot to lot traceability
Specifications

Specifications

White
ISO class 5 and above applications
Nonwoven Cleanroom Wipes
12 x 12 cm (9 x 9 in.)
Polyester Microfiber
Non-sterile
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit