Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Savant™ DNA SpeedVac™ Concentrator Kits Available on GSA/VA Contract for Federal Government customers only.

Save time and money with Thermo Scientific™ Savant™ DNA SpeedVac™ Concentrators Kits. Everything you need for quick drying and concentrating of small sample volumes.

Manufacturer:  Thermo Scientific™ DNA120115

 View more versions of this product

Catalog No. 20-548-130


  Check Availability
Add to cart

Specifications

Description

Save time and money with compact Thermo Scientific™ Savant™ DNA SpeedVac™ Concentrators Kits. These complete systems provide everything you need to quickly dry and concentrate small sample volumes in one fully integrated package, sold under one part number.

Operating Features

  • Consistent performance: designed to ensure consistent drying times and reproducible results
  • Simplicity: single program supports all DNA/RNA applications
  • User-friendly interface: with temperature/time settings
  • Three drying rates: ambient, ?43°C, ?65°C
  • Dual digital timers: independent control of heat/run times

Sample Protection

  • Dry at ambient temperatures or at very low heat to avoid sample denaturing
  • Oil-free diaphragm pump provides gentle transition from atmosphere to ultimate vacuum setting with no bumping

Safety Features

  • PTFE-coated chamber resists chemicals and cleans easily
  • Latched lid ensures user safety during operation
  • Unit and pump are virtually maintenance-free
  • Optional chemical traps and cartridges trap radioactivity and ammonia

Solvents and Solvent Combinations:

DNA120
  • PCR buffers (aqueous)
  • Water
  • Ethanol and water
DNA-120-OP
  • Ammonium hydroxide
  • Isopropanel
  • PCR buffers (aqueous)

ISS-110

  • PCR buffers (aqueous)
  • Ethanol and water
  • Isopropanol

DNA 120 SpeedVac Kit

  • Corrosion-resistant, PTFE coated chamber provides years of lasting service
  • Built-in, oil free vacuum pump provides maintenance-free operation
  • Integrated and compact design
  • Extremely quiet operation
  • Choice of three drying rates
  • Dual digital timers for independent control of heat and run times

DNA120OP SpeedVac Kit for oligos

  • All the features of the DNA 120
  • Plus external post trap to neutralize ammonium hydroxide, chemically resistant glass cover to contain solvents

High capacity DNA SpeedVac Kit ISS110

  • Built-in ETFE coated vacuum pump providing years of lasting service
  • Four liters, -50°C refrigerated vapor trap (integrated)
  • Choice of three drying rates

Includes:

DNA120

  • DNA concentrator with integrated pump
  • RD36 rotor for 36 x 1.6mL microtubes
DNA120-OP
  • DNA concentrator with integrated pump and glass cover
  • RD36 rotor for 36 x 1.5mL microtubes
  • ANT100 chemical trap
  • ANS121 ammonium neutralizing solution

Description & Specifications

Specifications

4L
PTFE
115V
31L/min. at 50Hz ; 36L/min. at 60Hz
DNA120; 115V, 60Hz
115V 60Hz
-55°C
7 torr (9 mbar)
60Hz
Concentrator
CE (230V only)
Accessories
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit